Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Actin-like 7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114300 Supplier Invitrogen™ Supplier No. PA5114300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYCKCGFAGL PRPTHKISTT VGKPYMETAK TGDNRKETFV GQELNNTNVH For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.

The protein encoded by this gene is a member of a family of actin-related proteins (ARPs) which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene (ACTL7A), and related gene, ACTL7B, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7A gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. The ACTL7A gene is expressed in a wide variety of adult tissues, however, its exact function is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin-like 7A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACTL7A
Gene Accession No. Q9Y615
Gene Alias actin like 7A; actin-like 7a; actin-like 7-alpha; actin-like protein 7A; actin-like Tact2; actin-like-7-alpha; ACTL7A; Arp7a; Tact2; T-actin 2; T-actin-2; testicular secretory protein Li 3; testis-specific actin-2
Gene Symbols ACTL7A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ACTL7A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10881
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.