Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Actin-like 7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544089 Supplier Invitrogen™ Supplier No. PA544089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KLITIGQERF RCSEMLFQPS LAGSTQPGLP ELTAACLGRC QDTGFKEEMA Sequence homology: Cow: 77%; Dog: 83%; Human: 100%; Mouse: 85%; Rat: 92%.

The protein encoded by this gene is a member of a family of actin-related proteins which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene, and related gene, ACTL7A, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7B gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. Unlike ACTL7A, the ACTL7B gene is expressed predominantly in the testis, however, its exact function is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin-like 7B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACTL7B
Gene Accession No. Q9Y614
Gene Alias actin like 7B; actin-like 7B; actin-like 7-beta; actin-like protein 7B; Actin-like-7-beta; ACTL7B; MGC151192; MGC151194; RP11-3J11.2; Tact1; t-actin 1; T-actin-1; testis tissue sperm-binding protein Li 43a; testis-specific actin-1; Unknown (protein for MGC:134108)
Gene Symbols ACTL7B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ACTL7B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10880
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.