Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Actin-like 7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554513
Description
Immunogen sequence: MATRNSPMPL GTAQGDPGEA GTRPGPDASL RDTGAATQLK MKPRKVHKIK AVIIDLGSQY CKCGYAGEPR PTYFISSTVG KRCPEAADAG DTRKWTL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.
The protein encoded by this gene is a member of a family of actin-related proteins which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene, and related gene, ACTL7A, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7B gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. Unlike ACTL7A, the ACTL7B gene is expressed predominantly in the testis, however, its exact function is not known.
Specifications
| Actin-like 7B | |
| Polyclonal | |
| Unconjugated | |
| ACTL7B | |
| actin like 7B; actin-like 7B; actin-like 7-beta; actin-like protein 7B; Actin-like-7-beta; ACTL7B; MGC151192; MGC151194; RP11-3J11.2; Tact1; t-actin 1; T-actin-1; testis tissue sperm-binding protein Li 43a; testis-specific actin-1; Unknown (protein for MGC:134108) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y614 | |
| ACTL7B | |
| Recombinant protein corresponding to Human Actin-like 7B. Recombinant protein control fragment (Product #RP-93001). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction