Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Mouse anti-Human,Eukaryotes, Unconjugated, Clone: MAb GEa (5-16 H1C8), MilliporeSigma™

Catalog No. MABT15332MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT15332MI 25 μL
MABT15331MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT15332MI Supplier MilliporeSigma Supplier No. MABT153325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin Monoclonal antibody specifically detects Actin in Human,Eukaryotes samples. It is validated for ELISA, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications ELISA, Western Blot
Classification Monoclonal
Clone MAb GEa (5-16 H1C8)
Conjugate Unconjugated
Gene Accession No. NP_850284.1
Gene Alias ACT1
Gene Symbols ACT1, AAC1, At2g37620, E13M22.12
Host Species Mouse
Immunogen Purified actin (ACT1) from Arabidopsis thaliana plant.
Molecular Weight of Antigen ∼44 kDa observed; 41.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Ligatrap
Quantity 25 μL
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 5-16 H1C8 is a mouse monoclonal antibody that detects Actin in a wide range of species.
Target Species Human, Eukaryote
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.