Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody

Catalog No. MS742RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS742RQ 12 mL Ready-to-Use
MS742S1 500 μL Concentrated
MS742S 1 mL Concentrated
MS742R7 7 mL Prediluted
MS742S0 100 μL Concentrated
MS742R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS742RQ Supplier Epredia™ Supplier No. MS742RQ

Detect muscle-specific actin in immunohistochemistry experiments with Epredia™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: HHF35

Isotype: IgG1/κ

Species Reactivity: Human, Rabbit and Rat. Others not known.

Epitope: Not determined

Immunogen: SDS extract of human myocardium

Positive Control: Muscle or sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Actin, Muscle Specific Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HHF35
Conjugate Unconjugated
Host Species Mouse
Immunogen SDS extract of human myocardium
Quantity 12 mL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.