Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Rabbit anti-Human, Mouse, Rat, Zebrafish, Clone: JF47-01, Novus Biologicals™
SDP

Catalog No. NBP267113 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267113 Supplier Novus Biologicals Supplier No. NBP267113

Rabbit Monoclonal Antibody

Actin Monoclonal specifically detects Actin in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Actin
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JF47-01
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Gene Symbols ACTA1
Host Species Rabbit
Immunogen Synthetic peptide within Human Actin aa 303-349/ 377. (SwissProt: P68133 Human; SwissProt: P68134 Mouse; SwissProt: P68136 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.