Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Actin, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABT1485MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABT1485MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT1485MI Supplier MilliporeSigma™ Supplier No. ABT1485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin (ACTA1), smooth muscle alpha-actin (ACTA2), cytoplasmic beta-actin (ACTB), cardiac muscle alpha-actin (ACTC1), cytoplasmic gamma-actin (ACTG1), and smooth muscle gamma-actin (ACTG2, a.k.a. ACTA3). Although actins show >90% overall sequence homology, isotypes do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic - and -actins are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt. In additon to serving as the basic building blocks of cytoskeletal microfilaments, actins are involved in multiple nuclear functions and adopt specific conformations that are important for their association with nuclear actin-binding proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ACTA1, ACTA, CFTD, NEM3, MPFD
Gene Accession No. P68133
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminal end sequence of mature skeletal muscle alpha-actin (ACTA1).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001091
Test Specificity This rabbit polyclonal antibody was raised against skeletal muscle alpha-actin (ACTA1) N-terminal end sequence that shares 80% sequence identity in the corresponding region with cardiac muscle alpha-actin (ACTC1), smooth muscle alpha-actin (ACTA2), and cytoplasmic gamma-actin (ACTG1). Immunogen sequence is not found in smooth muscle gamma-actin (ACTG2, a.k.a. ACTA3) and cytoplasmic beta-actin (ACTB).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.