Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
actin-related protein 2/3 complex subunit 1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | actin-related protein 2/3 complex subunit 1B |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
actin-related protein 2/3 complex subunit 1B Polyclonal antibody specifically detects actin-related protein 2/3 complex subunit 1B in Human samples. It is validated for Western BlotSpecifications
actin-related protein 2/3 complex subunit 1B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
actin related protein 2/3 complex, subunit 1B (41 kD), actin related protein 2/3 complex, subunit 1B, 41kDa, actin-related protein 2/3 complex subunit 1B, ARC41p41-ARCp40-ARC, Arp2/3 complex 41 kDa subunit, ARP2/3 protein complex subunit p41 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human actin-related protein 2/3 complex subunit 1B. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
10095 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title