Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin11 Antibody (mAb2345a), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253120R
Description
Actin11 Monoclonal specifically detects Actin11 in Plant samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Actin11 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Plant | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
mAb2345a | |
Western Blot | |
ACT11, Actin-11 | |
Purified Arabidopsis Actin11 | |
0.1 mL | |
Apoptosis, Cancer, Cellular Signaling | |
This mouse monoclonal antibody was generated against plant actin ACT11 and recognizes subclasses 2 and 3 of vegetative and subclasses 4 and 5 of pollen specific plant actins (arabidopsis ACT1, 3, 4, 7, 11, 12). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction