Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin11 Antibody (mAb2345a), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253120MFV450
Description
Actin11 Monoclonal antibody specifically detects Actin11 in Plant samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Actin11 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Plant | |
| Purified |
| Western Blot, Immunofluorescence | |
| mAb2345a | |
| Western Blot, Immunofluorescence | |
| ACT11, Actin-11 | |
| Purified Arabidopsis Actin11 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction