Learn More
Actin2 Antibody (mAb13a), Alexa Fluor™ 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP253122AF350
Catalog No. NP23122A30
Only null left
Add to Cart
Description
Description
Actin2 Monoclonal specifically detects Actin2 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Actin2
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
Mouse
Protein A purified
Apoptosis, Cancer, Cellular Signaling
This mouse monoclonal antibody was generated against plant actin ACT2 and recognizes subclasses 1 and 3 of vegetative plant actins (arabidopsis ACT2, 8, 11).
Store at 4°C in the dark.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
mAb13a
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ACT2, actin 2, Actin-2
Purified Arabidopsis Actin2
0.1 mL
Primary
Plant
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.