Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin2 Antibody (mAb13a), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253122F
Description
Actin2 Monoclonal specifically detects Actin2 in Plant samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Actin2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Plant | |
Purified |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
mAb13a | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ACT2, actin 2, Actin-2 | |
Purified Arabidopsis Actin2 | |
0.1 mL | |
Apoptosis, Cancer, Cellular Signaling | |
This mouse monoclonal antibody was generated against plant actin ACT2 and recognizes subclasses 1 and 3 of vegetative plant actins (arabidopsis ACT2, 8, 11). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction