Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin8 Antibody (mAbGPa), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253121B
Description
Actin8 Monoclonal specifically detects Actin8 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Actin8 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Plant | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
mAbGPa | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ACT8, Actin-8 | |
Purified Arabidopsis Actin8 | |
0.1 mL | |
Apoptosis, Cancer, Cellular Signaling | |
This mouse monoclonal antibody was generated against plant actin ACT8 and recognizes all tested subclasses of plant actins (arabidopsis ACT1, 2, 3, 4, 7, 8, 11, 12) and Dictyosetelium actin. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction