Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actinomycin D, 1 mg/mL, DMSO, PROMO, Thermo Scientific Chemicals

Catalog No. J67160XFPM
Click to view available options
Quantity:
1 mL

Applications

Actinomycin D has a wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells, and as an antibiotic in treatment of various malignant neoplasms, including Wilm's tumor and the sarcomas.

Solubility

Soluble in acetonitrilie or acetone at 10 mg/mL and in DMSO at a minimum of 1 mg/mL, slightly soluble in water.

Notes

Research Use only: Not intended for animal or human diagnostic or therapeutic use.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.44 g/mol
MDL Number MFCD00005033
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
Synonym Dactinomycin; Actinomycin C1
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Liquid
Quantity 1 mL
Solubility Information Soluble in acetonitrilie or acetone and in DMSO, slightly soluble in water.
Chemical Name or Material Actinomycin D
EINECSNumber 200-063-6
RTECSNumber AU1575000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.