Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ 97-120 Oil Impermeable Industrial Gloves

Catalog No. 19501056 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-501-056 11
19-501-053 8
19-501-054 9
19-501-055 10
19-501-057 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-501-056 Supplier Ansell Supplier No. 97120110

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

ActivArmr™ gloves protect hands in extreme applications with specialized oil impermeable technology, and impact resistant construction.

  • Oil Impermeable Barrier repels drilling fluids and oils to keep hands clean and dry
  • Mud-oil grip with cut and impact resistance protects hands, reduces fatigue, and increases productivity
  • Hi-vis gloves feature a reflective logo for enhanced visibility
  • Impact bumpers protect entire back of hand from medium-level impact injury
  • Ideal for rugged environments and extreme applications
  • All-day comfort that helps workers get the job done
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Back Material PVC
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Material Nylon, Spandex
Product Type Mechanic's Gloves
Size 11
EN388 Standard 4242BP
For Use With (Application) Oil and Gas Drilling
Glove Cuff Style Knit
Liner Material Aramid Fiber
Packaging Type Polybag
Series 97-120
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.