Learn More
Description
- HIVIZ™ design increases awareness of users for safety
- Double-dip nitrile coating provides grip and control when working with oils and other liquids
- HPPE seamless knit glove keeps hands cool and comfortable
- Double-dip nitrile coating defends against oil exposure and abrasion injuries
- Designed for dynamic environments which have impact and cut hazards present
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 5 |
| Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
| Construction | Knit |
| Coating Material | Nitrile |
| Color | Gray, Hi-vis Orange |
| Cut Resistance Rating | ANSI A4 |
| Product Type | Cut-resistant Gloves |
| Size | 11 |
| EN388 Standard | EN 388:4X43CP |
| For Use With (Application) | Engagement and release of drill string slips, drilling assembly and completion, handling jagged metal, production Services, drilling/extraction, heavy equipment servicing, transport and logistics |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
