Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ ActivArmr™ Cotton Liner Underglove

Catalog No. 17985330 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
11
12
19
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-985-330 8
17-985-329 7
17-985-331 9
17-985-332 19
17-985-333 11
17-985-334 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-985-330 Supplier Ansell™ Supplier No. 96005080
Add to Cart
Add to Cart

Engineered for sweat management to be worn under Rubber Insulated Gloves

  • Engineered for professionals, our cotton liner is designed to be worn under Rubber Insulated Gloves, providing sweat absorption and comfort.
  • Stay dry, reduce irritation, and enhance performance during long hours in demanding environments
  • Sweat management to be worn under Rubber Insulated Gloves
  • Enhanced comfort and breathability
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 0
ASTM F739 No
Chlorinated No
Coated Area Uncoated
Construction Knit
Accelerator Free No
Applications Industrial
Coating Material Uncoated
Color White
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Cotton
Powdered Powder Free
Product Type Industrial Gloves
Size 8
Sterility Non-sterile
Style Gunn
EN 421 No
FDA 510 (k) No
For Use With (Application) Liner Under an Unsupported Glove
For Use With (Equipment) Rubber Insulated Gloves
Gender Unisex
Glove Cuff Style Knit
Grip Design Smooth
Hem Color Yellow
Lining Lined
Packaging Type Polybag
Thumb Style Keystone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.