Learn More
Ansell™ ActivArmr™ Flame Retardant Cooling Vest
Catalog No. 19010054 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Large
2X-Large
4X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-054 Small
19-010-055 Large
19-010-056 2X-Large
19-010-057 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-010-054 Supplier Ansell™ Supplier No. 9761000S
Ansell™ ActivArmr™ Flame Retardant Cooling Vest prevents elevated body temperatures in people who work in hot environments or with extreme physical exertion.
  • Reduces heat stress
  • Comfortable to wear
  • Easy to dress
  • Recharges at room temperature when stored
  • Washable
  • Long life time
  • Available in a flame retardant version
TRUSTED_SUSTAINABILITY
Specifications
Material Marko™ FR Fabric
Product Type Cooling Vest
Size Small
Length (Metric) 84 to 100 cm
Length (English) Chest: 33 to 39 in.
Color Dark Blue
Cooling Time Up to 4 hours
For Use With (Application) Fire and Rescue; Mining; Police; Oil platforms; Chemical industry; Steel industry; Refineries; Nuclear plants; Paper mill; Military use; Diving