Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Activated Carbon Filter (2 Pack)
GREENER_CHOICE

Catalog No. 19830506 Shop All Justrite Safety Group Products
Change view
Click to view available options
No. per Pack:
2 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
19-830-506 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-830-506 Supplier Justrite™ Supplier No. 12849
Add to Cart
Add to Cart

For Use With: VaporTrap Solvent Waste Systems

  • For use with VaporTrap™ solvent waste kits

Specifications

Description Activated Carbon Filter
Diameter (English) Outer 1.9 in.
For Use With (Equipment) VaporTrap Solvent Waste Systems
Length (English) 7 in.
Material Charcoal-activated Carbon
No. per Pack 2 Pk.
Product Type Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.