Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Activated Carbon Filters
SDP

Catalog No. 22130530
Click to view available options
For Use With (Equipment):
Leica CV5030
Tissue Tek SCA Coverslipper

Efficient, cost-effective activated carbon filters for many commonly used laboratory instruments.

  • Filters contain high grade activated carbon which will absorb most organic and inorganic vapors including xylene, toluene, methanol, formalin, isopropyl alcohol and industrial denatured alcohol
  • These products are identical in size and shape to their comparable original manufacturer filters, and are an easy drop-in replacement
  • Change filters at manufacturer-recommended intervals to keep your laboratory and staff safe
  • 1 Ea.
  • Available for Leica CV5030 or Tissue Tek SCA

Specifications

Product Type Activated Carbon Filter
For Use With (Equipment) Tissue Tek SCA Coverslipper
Quantity 1 Ea.
Dimensions (L x W x H) 105 x 105 x 40 mm
Thickness (Metric) 40 mm
Width (Metric) 125 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.