Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activated charcoal, iodinated, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite,activated charcoal,norit,mineral,carbon-12,carbono,graphene,acticarbone,anthrasorb,carbosieve
5462310
CHEBI:27594
carbon
[C]
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite,activated charcoal,norit,mineral,carbon-12,carbono,graphene,acticarbone,anthrasorb,carbosieve
5462310
CHEBI:27594
carbon
[C]
Filter aid
Packaging Glass bottle
Color Black
Quantity 1kg
Merck Index 14, 1807
Solubility Information Solubility in water: insoluble
Formula Weight 12
Physical Form Thin Rods
Percent Purity 5%, exact strength on the certificate of analysis
Chemical Name or Material Activated charcoal

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 231-153-3