Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Active β2 Integrin (CD18) Mouse anti-Human, BUV 395, Clone: mAb 24 (also known as 24; m24; M24; mAb24), BD Horizon™

Catalog No. BDB567947 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567947 100 Tests
BDB567948 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB567947 Supplier BD Biosciences Supplier No. 567947
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Active β2 Integrin (CD18) Monoclonal antibody specifically detects Active β2 Integrin (CD18) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Active β2 Integrin (CD18)
Applications Flow Cytometry
Classification Monoclonal
Clone mAb 24 (also known as 24; m24; M24; mAb24)
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias ITGB2; Integrin beta-2; Integrin β2; LFA-1 β; LFA1b; beta 2 integrin; β2 integrin
Host Species Mouse
Immunogen Fibronectin-purified Human Monocytes
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.