Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Active β2 Integrin (CD18) Mouse anti-Human, R718, Clone: mAb 24 (also known as 24; m24; M24; mAb24), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567969
This item is not returnable.
View return policy
Description
Active β2 Integrin (CD18) Monoclonal antibody specifically detects Active β2 Integrin (CD18) in Human samples. It is validated for Flow cytometry (Routinely Tested)
Specifications
Active β2 Integrin (CD18) | |
Monoclonal | |
R718 | |
ITGB2; Integrin beta-2; Integrin β2; LFA-1 β; LFA1b; beta 2 integrin; β2 integrin | |
Fibronectin-purified Human Monocytes | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Flow Cytometry | |
mAb 24 (also known as 24; m24; M24; mAb24) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Purified |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction