Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International Active Anthrax Detect™ Plus Rapid Test
SDP

Catalog No. 21200115
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-115 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-115 Supplier InBios International Supplier No. AAP1
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Active Anthrax Detect™ Plus Rapid Test point-of-care diagnostic test for pulmonary anthrax is an in vitro immunochromatographic device for use as an aid in the diagnosis of inhalation anthrax.

  • For use in point-of-care settings by military and healthcare professionals
  • PPA (Relative Sensitivity): >98%
  • NPA (Relative Specificity): 100%
  • For use with venous whole blood or serum

Applications:

  • The Active Anthrax Detect™ Plus Rapid Test point-of-care diagnostic test for pulmonary anthrax is an in vitro immunochromatographic device for use as an aid in the diagnosis of inhalation anthrax. It provides visual and rapid qualitative detection of lethal factor of Bacillus anthracis (B. anthracis). The test can be used to test serum and venous whole blood (dipotassium EDTA, sodium citrate, and sodium heparin). The assay is indicated for testing samples from individuals who have signs and symptoms consistent with inhalation anthrax and a likelihood of exposure. This test is intended for use by military personnel, medical, and/or healthcare professionals only. The diagnosis of B. anthracis infection must be based on history, signs, symptoms, exposure likelihood, and additional laboratory evidence. A positive Active Anthrax Detect™ Plus Rapid Test result is presumptively diagnostic for B. anthracis infection. The definitive identification of B. anthracis from blood samples requires additional testing and confirmation procedures in consultation with public health or other authorities for whom reports are required. Testing should be performed and reported in accordance with current guidelines provided by the appropriate public health authorities. The level of lethal factor present in blood from individuals with early systemic infection is unknown. Negative results do not preclude infection with the biothreat microbial agents targeted by the device and should not be used as the sole basis for diagnosis, treatment, or other patient management decisions. This assay is for Rx use.

The distribution of in vitro diagnostic devices for Bacillus spp. detection is limited to laboratories that follow public health guidelines that address appropriate biosafety conditions, interpretation of test results, and coordination of findings with public health authorities.

This assay is not FDA-cleared or approved for testing blood or plasma donors.

Specifications

Content And Storage Store at Room Temperature (15°C to 30°C)
Detectable Analytes Antigen
Detection Method Rapid Test
Type Rapid Test
Immunoassay Kit Format Immunochromatographic Test
Quantity 25 Tests/Kit
Species B. anthracis
Analysis Time 20 minutes
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.