Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Active Caspase-3 Rabbit anti-Human, Mouse, BV650, Clone: C92-605.rMAb, BD Horizon™

Catalog No. BDB570179 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570179 100 Tests
BDB570182 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570179 Supplier BD Biosciences Supplier No. 570179
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Active Caspase-3 Monoclonal antibody specifically detects Active Caspase-3 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Active Caspase-3
Applications Flow Cytometry, Control
Classification Monoclonal
Clone C92-605.rMAb
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Apopain; CASP-3; CASP3; CC3; CPP-32; CPP32; Caspase 3; Yama
Host Species Rabbit
Immunogen Human Active Caspase-3 Fragment
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.