Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin A Antibody (105), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290669G
Description
Activin A Monoclonal specifically detects Activin A in Mouse samples. It is validated for ELISA.Specifications
Activin A | |
Monoclonal | |
DyLight 488 | |
activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Activin A (NP_032406.1; Met1-Ser424). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Stem Cell Signaling Pathway | |
3624 | |
Mouse | |
Purified |
ELISA | |
105 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human INHBA | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction