Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Activin A Recombinant Protein

Catalog No. p-7075587
Encompass_Preferred
Click to view available options
Quantity:
5 μg

Activin A Recombinant Human Protein

Human Activin A Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

Recombinant Human Activin A is a bioactive protein intended for use in cell culture applications. Activin A is involved in multiple biological processes including inflammation, neural development, and haematopoiesis.

  • High purity—no interference from other proteins or contaminants
  • High biological activity—more results with less protein
  • Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media

    High purity recombinant bioactive protein
    Recombinant Human Activin A is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

    High biological activity
    In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Activin A specific activity is determined by the dose dependent proliferation of mouse MPC-11 cells.

    Proven compatibility
    Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

    For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    For Use With (Application) Cell Culture
    Gene ID (Entrez) 3624
    Molecular Weight (g/mol) 26 kDa
    Purification Method Sequential Chromatography
    Quantity 5 μg
    Gene Alias Activin A
    Species Human
    Content And Storage Store in freezer (-5 to -30°C).
    Endotoxin Level < 0.1 ng/μg
    Shipping Condition Approved for shipment on Wet or Dry Ice
    Expression System Baculovirus
    Protein Family Growth Factors & Receptors
    Protein Form Recombinant, Ligand
    Form Lyophilized
    Protein Subtype Activins
    Research Category Neurobiology, Stem Cell Research, Differentiation
    Product Line Gibco™
    Purity or Quality Grade 95 %
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.