Learn More
Invitrogen™ Activin A Recombinant Rabbit Monoclonal Antibody (1X6N9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542875
Description
Positive test controls include: HeLa, Mouse uterus, Rat testis. The target is usually found in the following locations: Secreted. Immunogen sequence: MPLLWLRGFL LASCWIIVRS SPTPGSEGHS AAPDCPSCAL AALPKDVPNS QPEMVEAVKK HILNMLHLKK RPDVTQPVPK AALLNAIRKL HVGKVGENGY.
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
Specifications
Activin A | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08476, P18331, Q04998 | |
Inhba | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Inhibin beta A (INHBA) (P08476). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
1X6N9 | |
Unconjugated | |
Inhba | |
activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
16323, 29200, 3624 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.