Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin C/Inhibin beta C Antibody (CL6916), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Activin C/Inhibin beta C |
---|---|
Clone | CL6916 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.Specifications
Activin C/Inhibin beta C | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
inhibin beta C chain, IHBC, inhibin, beta C | |
INHBC | |
IgG2b | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL6916 | |
Monoclonal | |
Purified | |
Cytokine Research | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
3626.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TPRAGGQCPACGGPTLELESQRELLLDLAKRSILDKLHLTQRPTLNRPVSRAALRTALQHLHGVPQGALLEDNREQECEI | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title