Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin C/Inhibin beta C Antibody (MM0076-4N17), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212011H
Description
Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.Specifications
Activin C/Inhibin beta C | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
INHBC | |
Human recombinant Activin C/Inhibin beta C | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0076-4N17 | |
Western Blot | |
inhibin beta C chain, IHBC, inhibin, beta C | |
Mouse | |
Protein G purified | |
Cytokine Research | |
3626 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction