Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin RIB/ALK-4 Antibody (544310), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2221A
Description
Activin RIB/ALK-4 Monoclonal antibody specifically detects Activin RIB/ALK-4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Activin RIB/ALK-4 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
544310 | |
Flow Cytometry, CyTOF-ready | |
activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ACTRIB, ACVRLK4ACTR-IB, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B | |
Mouse myeloma cell line NS0-derived recombinant human Activin RIB/ALK-4, Ser24-Glu126, Accession # P36896 | |
0.1 mL | |
Protein Kinase | |
91 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction