Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin RIIA Antibody (SR2253), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32229750UL
Description
Activin RIIA Monoclonal antibody specifically detects Activin RIIA in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Activin RIIA | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2253 | |
Western Blot 1:500-1:2000 | |
activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ACTRII, ACTRIIA, ACVR2ACTR-IIA, EC 2.7.11, EC 2.7.11.30 | |
A synthesized peptide derived from human Activin RIIA (Uniprot #: P27037) | |
50 μg | |
Cancer | |
92 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction