Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin RIIB Antibody (307), FITC

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290465F
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Mouse samples. It is validated for ELISA.Specifications
Activin RIIB | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human ACVR2B | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
307 | |
ELISA | |
activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ACTRIIB, ACTR-IIB, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Activin RIIB extracellular domain (NP_031423.1; Met 1-Thr 134). | |
0.1 mL | |
Protein Kinase | |
93 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction