Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTL7A Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ACTL7A |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ACTL7A Polyclonal specifically detects ACTL7A in Mouse samples. It is validated for Western Blot.Specifications
ACTL7A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Chromatin Research | |
PBS buffer, 2% sucrose | |
10881 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
actin-like 7A, actin-like 7-alpha, actin-like protein 7A, actin-like-7-alpha | |
The immunogen is a synthetic peptide directed towards the middle region of Mouse ACTL7A (NP_033741). Peptide sequence VVPIYEGYPLPSITGRLDYAGSDLTTYLMNLMNNSGKHFSEDHLGIVEDI | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title