Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTL7B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP186973
Description
ACTL7B Polyclonal specifically detects ACTL7B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ACTL7B | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
actin-like 7B, actin-like 7-beta, actin-like protein 7B, actin-like-7-beta, Tact1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ACTL7B | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MATRNSPMPLGTAQGDPGEAGTRPGPDASLRDTGAATQLKMKPRKVHKIKAVIIDLGSQYCKCGYAGEPRPTYFISSTVGKRCPEAADAGDTRKWTL | |
0.1 mL | |
Chromatin Research | |
10880 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction