Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTL9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555380
Description
Immunogen sequence: GHGVTYTVPV FQGYNLLHAT ERLDLAGNHL TAFLAEMLLQ AGLPLGQQDL DLVENIKHHY CYVASDFQKE QARPEQEYKR TLKLPDGRTV TLGKELFQC Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.

Specifications
ACTL9 | |
Polyclonal | |
Unconjugated | |
ACTL9 | |
1700029I08Rik; actin like 9; actin-like 7A; actin-like 7-alpha; actin-like 9; actin-like protein 9; ACTL7C; ACTL9; AV259599; HSD21; testicular tissue protein Li 15; testicular tissue protein Li 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
284382 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TC94 | |
ACTL9 | |
Recombinant protein corresponding to Human ACTL9. Recombinant protein control fragment (Product #RP-92491). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction