Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTL9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117695
Description
This antibody has been tested in direct-ELISA.

Specifications
ACTL9 | |
Polyclonal | |
Unconjugated | |
ACTL9 | |
1700029I08Rik; actin like 9; actin-like 7A; actin-like 7-alpha; actin-like 9; actin-like protein 9; ACTL7C; ACTL9; AV259599; HSD21; testicular tissue protein Li 15; testicular tissue protein Li 9 | |
Rabbit | |
Protein G | |
RUO | |
284382, 69481 | |
-20°C or -80°C if preferred | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.25 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8CG27, Q8TC94 | |
ACTL9 | |
Recombinant Human Actin-like protein 9 protein (1-160aa). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction