Learn More
Invitrogen™ alpha Actinin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578720
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HT1080 whole cell, rat skeletal muscle tissue, rat skeletal muscle tissue, mouse skeletal muscle tissue, mouse skeletal muscle tissue. IHC: mouse skeletal muscle tissue, rat skeletal muscle tissue, human lung cancer tissue. Flow: WISH cell.
Alpha-actinin-3, also known as alpha-actinin skeletal muscle isoform 3 or F-actin cross-linking protein, is a protein that in humans is encoded by the ACTN3 gene. This gene encodes a member of the alpha-actin binding protein gene family. The encoded protein is primarily expressed in skeletal muscle and functions as a structural component of sarcomeric Z line. This protein is involved in crosslinking actin containing thin filaments. An allelic polymorphism in this gene results in both coding and non-coding variants; the reference genome represents the coding allele.
Specifications
alpha Actinin 3 | |
Polyclonal | |
Unconjugated | |
Actn3 | |
actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 3; Actinin-alpha3; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; alpha-Actinin3; Alpha-actinin-3; alphaActinin-3; F-actin cross-linking protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11474, 171009, 89 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O88990, Q08043 | |
Actn3 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human ACTN3 (574-617aa EADRERGAIMGIQGEIQKICQTYGLRPCSTNPYITLSPQDINT K). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.