Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTR10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544739 Supplier Invitrogen™ Supplier No. PA544739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SLIQCPIDTR KQLAENLVVI GGTSMLPGFL HRLLAEIRYL VEKPKYKKAL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

ACTR10is a protein coding gene. Gene Ontology (GO) annotations related to this gene include neutrophil degranulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTR10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACTR10
Gene Accession No. Q9NZ32
Gene Alias Act11; actin related protein 10; actin related protein 10 homolog; actin-related protein 10; actin-related protein 10 homolog; actin-related protein 10 homolog (S. cerevisiae); actin-related protein 10-like protein; actin-related protein 11; Actr10; ACTR11; Arp10; ARP10 actin related protein 10 homolog; ARP10 actin-related protein 10; ARP10 actin-related protein 10 homolog; Arp11; hARP11; hypothetical protein LOC511083; LOW QUALITY PROTEIN: actin-related protein 10; zgc:55635
Gene Symbols ACTR10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ACTR10.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55860
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.