Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTR10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590774 Supplier Invitrogen™ Supplier No. PA590774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDLKRGLKIQ AAKFNIDGNN ERPSPPPNVD YPLDGEKILH ILGSIRDSVV EILFEQDNEE QSVATLILDS LIQCPIDTRK QLAENLVVIG GTSMLPGFLH RLLAEIRYLV EKPKYKKALG TKTFRIHTPP AKANCVAWLG GAIFGALQDI LGSRSVSKEY YNQTGRIPDW CSLNNPPLEM MFDVGKTQPP LMKRAFSTEK; Positive Samples: SKOV3, 293T, Mouse heart, Mouse kidney, Mouse lung; Cellular Location: Cytoplasm, cytoskeleton.

ACTR10is a protein coding gene. Gene Ontology (GO) annotations related to this gene include neutrophil degranulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTR10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ACTR10
Gene Accession No. Q9NZ32, Q9QZB7
Gene Alias Act11; actin related protein 10; actin related protein 10 homolog; actin-related protein 10; actin-related protein 10 homolog; actin-related protein 10 homolog (S. cerevisiae); actin-related protein 10-like protein; actin-related protein 11; Actr10; ACTR11; Arp10; ARP10 actin related protein 10 homolog; ARP10 actin-related protein 10; ARP10 actin-related protein 10 homolog; Arp11; hARP11; hypothetical protein LOC511083; LOW QUALITY PROTEIN: actin-related protein 10; zgc:55635
Gene Symbols ACTR10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 218-417 of human ACTR10 (NP_0609471).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299121, 55860, 56444
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.