Learn More
Invitrogen™ ACTR1A Monoclonal Antibody (3E5)

Description
Peptide Sequence: LVFAIQKSDM DLRRTLFSNI VLSGGSTLFK GFGDRLLSEV KKLAPKDVKI RISAPQERLY STWIGGSILA SLDTFKKMWV SKKEYEEDGA RSIHRKT.
Specifications
Specifications
| Antigen | ACTR1A |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3E5 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ACTR1A |
| Gene Accession No. | P61163 |
| Gene Alias | actin related protein 1A; actin-related protein 1 alpha; actin-related protein 1 homolog A; actin-RPV; ACTR1A; actr1a {ECO:0000250; alpha-Arp1; alpha-centractin; ARP1; ARP1 actin related protein 1 homolog A; ARP1 actin-related protein 1 homolog A, centractin alpha; ARP1 actin-related protein 1 homolog A, centractin alpha (yeast); ARP1 actin-related protein 1A, centractin alpha; Arp1 centractin alpha; Arp1 contractin alpha; Arp1A; centractin; Centrosome-associated actin homolog; CTRN1; FLJ52695; FLJ52800; FLJ55002; UniProtKB:P61163}; yeast homolog A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.