Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTR1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593024 Supplier Invitrogen™ Supplier No. PA593024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Mouse skeletal muscle, Mouse spleen, Mouse lung, Rat brain; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center.

This gene encodes a 42.6 kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 8-13 copies per dynactin molecule, and is the most abundant molecule in the dynactin complex. It is an actin-related protein, and is approximately 60% identical at the amino acid level to conventional actin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTR1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACTR1A
Gene Accession No. P61163, P61164, P85515
Gene Alias actin related protein 1A; actin-related protein 1 alpha; actin-related protein 1 homolog A; actin-RPV; ACTR1A; actr1a {ECO:0000250; alpha-Arp1; alpha-centractin; ARP1; ARP1 actin related protein 1 homolog A; ARP1 actin-related protein 1 homolog A, centractin alpha; ARP1 actin-related protein 1 homolog A, centractin alpha (yeast); ARP1 actin-related protein 1A, centractin alpha; Arp1 centractin alpha; Arp1 contractin alpha; Arp1A; centractin; Centrosome-associated actin homolog; CTRN1; FLJ52695; FLJ52800; FLJ55002; UniProtKB:P61163}; yeast homolog A
Gene Symbols ACTR1A
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10121, 294010, 54130
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.