Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTR1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116639 Supplier Invitrogen™ Supplier No. PA5116639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse thymus, Mouse brain, Mouse heart, Rat lung Immunogen sequence: MESYDIIANQ PVVIDNGSGV IKAGFAGDQI PKYCFPNYVG RPKHMRVMAG ALEGDLFIGP KAEEHRGLLT IRYPMEHGVV RDWNDMERIW QYVYSKDQLQ TFSEE.

This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTR1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Actr1b
Gene Accession No. P42025, Q8R5C5
Gene Alias 2310066K23Rik; AA960180; Actin-related protein 1B; Actr1b; AI851923; ARP1; ARP1 actin-related protein 1 homolog B, centractin beta; ARP1 actin-related protein 1 homolog B, centractin beta (yeast); ARP1 actin-related protein 1B, centractin beta; Arp1b; Beta-centractin; centractin beta; CTRN2; PC3; yeast homolog B
Gene Symbols Actr1b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human ACTR1B (NP_0057261).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10120, 226977, 316333
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.