Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR3 Antibody (7E5), Novus Biologicals™
SDP

Catalog No. NB297926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297926 50 μL
NB297927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297926 Supplier Novus Biologicals Supplier No. NBP32652550UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ACTR3 Monoclonal antibody specifically detects ACTR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen ACTR3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7E5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3
Host Species Rabbit
Immunogen A synthesized peptide derived from Human ACTR3 [UniProt P61158]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10096
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.