Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTR3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297009V
Description
ACTR3 Polyclonal specifically detects ACTR3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ACTR3 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
10096 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
DyLight 405 | |
actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3 | |
Produced in rabbits immunized with purified, recombinant ACTR3 (NP_005712.1; Ala2-Ser418). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction