Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTR3 Mouse anti-Human, Clone:3G2G1, Abnova™
Mouse monoclonal antibody raised against partial recombinant human ACTR3.
Supplier: Abnova Corporation MAB17986
Description
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion. [provided by RefSeq]Specifications
ACTR3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Immunofluorescence (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
ACTR3 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3G2G1 | |
ARP3 actin-related protein 3 homolog (yeast) | |
In PBS (0.05% sodium azide) | |
ARP3 | |
Recombinant protein corresponding to amino acids 287-418 of human ACTR3. | |
10096 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction