Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTR3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596824 Supplier Invitrogen™ Supplier No. PA596824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CIKHIPIAGR DITYFIQQLL REREVGIPPE QSLETAKAIK EKYCYICPDI VKEFAKYDVD PRKWIKQYTG INAINQKKFV IDVGYERFLG PEIFFHPEFA NPDFMESISD VVDEVIQNCP IDVRRPLYKN VVLSGGSTMF RDFGRRLQRD LKRVVDARLR LSEELSGGRI KPKPVEVQVV THHMQRYAVW FGGSMLASTP EFFQVCHTKK DYEEYGPSIC RHNPVFGVMS; Positive Samples: HeLa, NCI-H125, Mouse brain; Cellular Location: Cell projection, Cytoplasm, cytoskeleton.

ACTR3B plays a role in the organization of the actin cytoskeleton.ACTR3B may function as ATP-binding component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks. ACTR3B may decrease the metastatic potential of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTR3B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACTR3B
Gene Accession No. Q641P0, Q9P1U1
Gene Alias 9630005C02; actin related protein 3B; actin-like protein 3B; actin-related protein 3B; actin-related protein 3-beta; actin-related protein Arp11; actin-related protein ARP4; ACTR3B; ARP11; ARP3 actin related protein 3 homolog B; ARP3 actin-related protein 3 homolog B; ARP3 actin-related protein 3 homolog B (yeast); ARP3 actin-related protein 3B; Arp3b; Arp3beta; ARP3-beta; ARP4; AW047569; DKFZp686O24114; RGD1565759; zgc:158823
Gene Symbols ACTR3B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 189-418 of human ACTR3B (NP_0651781).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242894, 57180
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.