Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560075 Supplier Invitrogen™ Supplier No. PA560075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLIISSGYQC THVLPILEGR LDAKNCKRIN LGGSQAAGYL QRLLQLKYPG HLAAITLSRM EEILHEHSYI AEDYVEELHK WRCPDYYENN VHKMQLPFSS KLL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

ACTR5 may be involved in transcription regulation via its interaction with the INO80 complex, a chromatin remodeling complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTR5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Actr5
Gene Accession No. Q9H9F9
Gene Alias AA545173; Actin-related protein 5; ACTR5; ARP5; ARP5 actin-related protein 5; ARP5 actin-related protein 5 homolog; ARP5 actin-related protein 5 homolog (yeast); B430109J19Rik; hARP5; I79_002649; INO80 complex subunit M; INO80M; sarcoma antigen NY-SAR-16
Gene Symbols Actr5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACTR5. Recombinant protein control fragment (Product #RP-99346).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.