Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTR5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100394
Description
Antibody detects endogenous levels of total Arp5/ACTR5.
ACTR5 may be involved in transcription regulation via its interaction with the INO80 complex, a chromatin remodeling complex.
Specifications
ACTR5 | |
Polyclonal | |
Unconjugated | |
Actr5 | |
AA545173; Actin-related protein 5; ACTR5; ARP5; ARP5 actin-related protein 5; ARP5 actin-related protein 5 homolog; ARP5 actin-related protein 5 homolog (yeast); B430109J19Rik; hARP5; I79_002649; INO80 complex subunit M; INO80M; sarcoma antigen NY-SAR-16 | |
Rabbit | |
Affinity chromatography | |
RUO | |
109275, 362258, 79913 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q80US4, Q9H9F9 | |
Actr5 | |
A synthesized peptide derived from human ACTR5(Accession Q9H9F9), corresponding to amino acid residues H249-Q299. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction