Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTRT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531691
Description
Recommended positive controls: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein related to the cytoskeletal protein beta-actin. This protein is a major component of the calyx in the perinuclear theca of mammalian sperm heads, and it therefore likely functions in spermatid formation. This gene is intronless and is similar to a related gene located on chromosome 1. A related pseudogene has also been identified approximately 75 kb downstream of this gene on chromosome X.
Specifications
| ACTRT1 | |
| Polyclonal | |
| Unconjugated | |
| ACTRT1 | |
| actin related protein T1; actin-related protein T1; ACTRT1; AIP1; ARIP1; ARPT1; ARP-T1; HSD27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 139741 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.93 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8TDG2 | |
| ACTRT1 | |
| Recombinant protein encompassing a sequence within the center region of human ACTRT1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction